Would you like to log in again?
PROSAT® Sterile Presaturated Nonwoven Wipers, Contec®
Supplier: Contec
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
PROSAT®
PSPP0039 PS-911EB PS-840IR PSC20005 PSC20005 PS-911ETOH PSPP0039 PS-911EB
10024-280CS 759.14 USD InStock
10024-280 CA11007-730 CA33503-136 10014-422 CA11011-780 CA68310-404 33503-136 68310-404
PROSAT® Sterile Presaturated Nonwoven Wipers, Contec®
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
Meltblown polypropylene wipes provide a consistent release of solvent to thoroughly remove surface contaminants in critical environments. PROSAT® presaturated wipes are exceptionally clean and also free from additives of any kind. This particular meltblown polypropylene contains very low levels of sodium and other ions.
  • Suitable for Grade A/B or ISO Class 5 environments
  • Validated sterile per gamma irradiation
  • Low in particles and fiber; ideal for product contact areas
  • Microsized filaments provide exceptional particle removal
  • Especially suitable for the metered release of 70% alcohol
  • Reduces VOC emissions and solvent use
  • Resealable pouch preserves cleanliness and solvent saturation levels
PROSAT® meltblown polypropylene wipes are ideal for wiping articles prior to pass through, cleaning process tools and other equipment, and general surface cleaning. Meltblown polypropylene fabric is exceptionally clean and free from additives.
Presaturated wipes ensure consistent saturation of each wipe independent of operator. The wipes are provided in convenient and easy to use peel and reseal pouches.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty