Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Stir Mantles, Large Capacity, Glas-Col®

Supplier: Glas-Col

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

100DEMS117 100DEMS121 100DEMS122 100DEMS120 100DEMS119 100DEMS116 100DEMS118
33725-082EA 3612.5 USD InStock
33725-082 33725-070 33725-080 33725-078 33725-076 33725-074 33725-072
Stir Mantles, Large Capacity, Glas-Col®
Heaters Heating Mantles
Heating and stirring in one unit.

  • 1/50 HP motor
  • Variable speed control
  • Large capacity
  • Total mixing and heating management
  • For use with spherical flask

This system has the power to mix larger volumes in a flask without an overhead stirrer. Powerful motor/magnet combination generates speed range of 100-1850 rpm. Variable motor speed control has detachable cord. Mantle is aluminum housed with fabric interior to softly nestle glassware. Ideal for temperatures up to 400 degrees C. The mantle is mounted on 4" feet to allow bench clearance for the stir motor and air flow.

Ordering information: mantle, stir control and 3/4 x 3" long stirring bar.

Delivery information: 7-10 days after order.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty