Would you like to log in again?
TAPE ALUM FOIL 2MIL 18YD EMI SHIELDING
Supplier: R S HUGHES
3M* Aluminum Foil Tape 5.1cm^ 3M* Aluminum Foil Tape, Dead-soft aluminum foil tape, Foil Thickness: 2 mil, Flame Retardant: Flame Retardant, Resistant Properties: Heat-Reflective, Width: 2 in, Length: 18 yd, Total Thickness: 2 mil, Tensile Strength: 20 lb/in, Application: EMI Shielding
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
TAPE ALUM FOIL 2MIL 18YD EMI SHIELDING 33725-531 1170 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty