Would you like to log in again?
TAPE FOIL 1 INCH CS36
Supplier: R S HUGHES
3M* Aluminum Foil Tape^ Width: 2.5cm. Length: 54.9m. Dead-soft aluminum foil tape. Unlinered version. Can be easily die-cut into special sizes or shapes. Masks sensitive components to protect from damage during paint stripping or reflect and dissipate heat.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
TAPE FOIL 1 INCH CS36 33725-568 425 Case of 36 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty