Would you like to log in again?
Mantle/Controller Package, Series TM, Glas-Col®
Supplier: Glas-Col
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
100DTM106PL 100DTM112PL 100DTM102PL 100DTM104PL 100DTM114PL 100DTM108PL 100DTM110PL
33752-442EA 1026.39 USD InStock
33752-442 33752-420 33752-464 33752-486 33752-500 33752-522 33752-409
Mantle/Controller Package, Series TM, Glas-Col®
Heaters Heating Mantles
Glas-Col® offers TM series mantles and PowrTrol power control (see 33797-414 series) together in a special package
CSA certified.
The compact control is a solid-state, proportional voltage manual control that provides precise control of the output range, adjustable from 3–100%. Control has a lighted on/off power switch, three-wire load receptacle, and three-wire cord with molded plug. Series TM mantles feature an aluminum exterior with fabric interior, units up to 600W, 120V, 5A. Mantles withstand an internal operating temperature of 450°C (840°F).
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty