Would you like to log in again?
Mantle for Resin Reaction Flasks, Series TM, Glas-Col®
Supplier: Glas-Col
100BTM578 100BTM570 100BTM572 100BTM580 100BTM576
33754-040EA 584.04 USD InStock
33754-040 33754-084 33754-109 33754-029 33754-120
Mantle for Resin Reaction Flasks, Series TM, Glas-Col®
Heaters Heating Mantles
Series TM resin reaction flask mantle provides even heat distribution to prevent scorching of clear resinous material
CSA certified.
One-circuit heat input simplifies temperature control. Aluminum exterior with fabric interior to softly nest glass vessels. Withstands 450°C (840°F) internal operating temperature. Detachable 1.2m (4') long, 3-wire cord and locking connector. For 115V operation. For 230V units, contact your Avantor representative.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty