Would you like to log in again?
Mantle for Griffin Beakers, Series TM, Glas-Col®
Supplier: Glas-Col
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
100BTM610 100BTM612 100BTM620 100BTM618 100BTM616 100BTM614
33758-100EA 694.64 USD InStock
33758-100 33758-121 33758-020 33758-041 33758-085 33758-063
Mantle for Griffin Beakers, Series TM, Glas-Col®
Heaters Heating Mantles
Series TM Griffin beaker mantle has a resilient and pliable interior to cradle fragile beakers
CSA certified.
Heat is concentrated at the bottom so even small liquid volumes can be heated efficiently. Mantles fit KIMAX® brand beakers from series 89001-054. Detachable 1.2m (4') long, 3-wire cord and locking connector. For 115V operation. Also available in 230V.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty