Would you like to log in again?
Heating Mantle Top, Top Only, Glas-Col®
Supplier: Glas-Col
100AMO1163 100AMO1123 100AMO1083 100AMO1143 100AMO1063
33779-380EA 1375.87 USD InStock
33779-380 33779-369 33779-389 33779-289 33779-295
Heating Mantle Top, Top Only, Glas-Col®
Heaters Heating Mantles
Prevents vapor condensation in top half of flask during distillation.
  • Protects flask during process
  • Laced side opening
  • Elliptical hole accomodates 1, 2 and 3 neck flasks
Our heated top mantle covers the upper half of a flask during distillation of high boiling materials. The tops are designed for use with our M, TM or STM mantles. They protect the flask from adverse conditions which would disturb the reflux ration during fractionation.
Delivery information: 7-10 days after order.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty