Would you like to log in again?
BioNeb® Nebulizing Cell Disruption Systems, Glas-Col®
Supplier: Glas-Col
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
BioNeb®
105ABN3015 105ABN3010 105ABN3025
33786-504EA 16753.62 USD InStock
33786-504 33786-502 33786-500
BioNeb® Nebulizing Cell Disruption Systems, Glas-Col®
Homogenizers Ultrasonic Cell Disruptors
These efficient and versatile systems can be used to gently open cells or completely disrupt them
Greater than 90% disruption of many types of cells can be achieved in only one or two passes. Fast, efficient process disrupts even the toughest cells in seconds. System is controlled by regulation of helium or nitrogen gas pressure, allowing for precise adjustments. Each system has single-pass batch operation, single-pass continuous flow operation, and continuous recycling operation modes.
BioNeb® systems do not require electricity, pressure chambers, or pumps. Unlike sonication and high-pressure liquid shear cell disruption systems, these systems do not require an expensive cooling system. BioNeb® cell disruption systems do not generate heat, minimizing sample damage. Easy to use, clean, and move.
Systems enable the gentle opening of cells without damaging internal structures, as necessary for organelle isolation, and the thorough breakage of intact cells, as necessary for preparation of a cell extract. Systems can also shear large biomolecules such as DNA to a desired size.
Ordering information: Supplied with cylinder for the appropriate sample volumes, universal positioner with base plate, flowmeter, and tubing.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty