Would you like to log in again?
Hemispherical Mantles, Series O, Glas-Col®
Supplier: Glas-Col
100AO408 0616700227 100AO410 100AO406 100AO398 100AO402 100AO396 100AO414 0616700220 100AO416 100AO412 100AO394 0616700229 100AO404 100AO400 0616700228
33787-108EA 373.85 USD InStock
33787-108 33787-061 33787-083 33787-040 33787-163 33787-130 33787-185 33787-141 33787-120 33787-132 33787-221 33787-265 33787-134 33787-200 33787-136 33787-028
Hemispherical Mantles, Series O, Glas-Col®
Heaters Heating Mantles
These hemispherical-style mantles cover only the bottom half of the flask, allowing full view of contents
Constructed of flexible fabrics for effective heating and protection of glass vessels. Choose from glass fabric exterior or silicone-impregnated glass fabric exterior. Mantles accommodate single and multi-neck flasks and adapt easily to unusual shapes. Withstand 450°C (840°F) internal operating temperature.
Ordering information: The use of an extension support is required (see 33793-018 series). Mantles include a separable 1.2m (4') long two-wire cord and locking connector. Mantles 33787-130, -132, -134, and -136 include a separable 1.2m (4') long three-wire cord with locking connector and grounding plate to ground the cord.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty