Would you like to log in again?
Stir Mantles, Glas-Col®
Supplier: Glas-Col
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
100DTEM102 100DTEM112 100DTEM108 100DTEM104 100DTEM106
33787-304EA 1122.92 USD InStock
33787-304 33787-306 33787-308 33787-312 33787-302
Stir Mantles, Glas-Col®
Heaters Heating Mantles
Heating and stirring in one unit.
  • Electromagnetic
  • Independent heating and stirring
  • Rigid housing for strength and support
  • For use with spherical flask
This system has the power to mix small volumes in a flask without an overhead stirrer. Electromagnetic speed range of 50-750 rpm. Mantle section is aluminum housed with fabric interior liner to softly nestle glassware. Ideal for temperatures up to 400 degrees C.
Delivery information: 7-10 days after order.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty