Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nutating 3D Shaker, 240 V

Supplier: Glas-Col
099A NSM24-2H
33787-632EA 13611.8 USD InStock
33787-632
Nutating 3D Shaker, 240 V
Shakers and Mixers Rotating/Rocking Shakers
Heavy duty shaker to handle heavier loads and provide greater durability

  • 240 V model
  • Bench top shaking
  • Three dimensional motion
  • Handles loads up to 40 pounds
  • Accomodates a variety of holders
  • Touch screen interface

This Nutating shaker gives you 3D shaking on your bench top. Ideal for environmental and extraction work using separatory funnels. This model is the base equipped with four 2 liter separatory funnel holders.

Ordering information: Four 2L Sep. Funnel Holders/Base.

Delivery information: 7-10 days after order.

Caution: 240 V model. Corresponding model with voltage suitable for use in US (33787-640) is also available, please see separate product page.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty