Would you like to log in again?
Mantle Extension Support, Series O, Glas-Col®
Supplier: Glas-Col
100EO535 100EO530 100EO524 100EO525 100EO522
33793-018EA 133.11 USD InStock
33793-018 33793-086 33793-064 33793-020 33793-042
Mantle Extension Support, Series O, Glas-Col®
Supports Flask Supports
Table-top support holds hemispherical and spherical Series O mantles up to 1L capacity
Steel support, with its fabric strap basket, is strong, handy and durable. Attaches easily to stable ring stands or distillation racks. Gray finish.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty