Would you like to log in again?
Three-Position and Six-Position Combination Heating Mantles, Glas-Col®
Supplier: Glas-Col
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
100DRX12712 100DRJ30012 100DRX100024 100DRJ50212E 100DRX30412E 100DRJ12724E 100DRJ80012 100DRX50212E 100DRX65012 100DRX30024E 100DRX50024 100DRJ12512E 100DRJ30012E 100DRJ50012E 100DRJ65024 100DRX65024 100DRJ12512 100DRX50012 100DRX65024E 104ARL4312 104ARL4324 100DRJ50224E 100DRX50024E 100DRJ80012E 100DRJ80024E 100DRX12512E 100DRJ65024E 100DRJ30412E 100DRX80012E 100DRJ30424 100DRJ50212 104ARL3612 100DRJ50012 100DRJ65012 100DRJ100024 100DRX50012E 100D RX100012E 100DRX12712E 100D RX100024E 104ARL3324 100DRJ12724 100DRJ30024 104ARL3624 100DRJ30424E 100DRX30424 100DRJ12524 100DRJ80024 100D RJ100024E 100DRJ30024E 100DRJ50024E 100DRX80024 100DRX65012E 100DRX30012E 100DRJ12712E 100DRX30012 100DRX50212 104ARL3312 100DRX80012 100DRX30424E 100DRX100012 100DRJ12524E 100DRX12512 100DRX30412 100DRJ50024 100DRX50224E 100DRX80024E 104ARL4612 100DRX30024 100DRJ30412 100DRJ50224 100DRX50224 104ARL4624
33795-256EA 3382.94 USD InStock
33795-256 33795-258 33795-292 33795-294 33795-250 33795-252 33795-296 33795-298 33795-254 33795-330 33796-022 33796-020 33796-064 33796-062 33796-060 33795-290 33796-028 33796-026 33796-024 33795-322 33795-288 33795-324 33795-326 33795-328 33795-280 33795-282 33795-284 33795-320 33795-286 33796-032 33796-030 33795-318 33796-038 33796-036 33796-034 33795-278 33795-310 33795-312 33795-314 33795-316 33795-270 33795-272 33795-274 33795-276 33796-044 33796-000 33796-042 33796-040 33796-008 33795-308 33796-006 33796-004 33796-048 33796-046 33795-300 33795-266 33795-302 33795-268 33795-304 33795-306 33795-260 33795-262 33796-010 33796-054 33796-052 33796-050 33796-018 33796-016 33796-014 33796-058 33796-056 33796-012
Three-Position and Six-Position Combination Heating Mantles, Glas-Col®
Heaters Heating Mantles
Ideal for repetitive extracting, refluxing and distilling laboratory applications.
Low-profile combination mantle utilizes convenient heating element container system that allows quick and easy replacement of a failed element and eliminates the need to tear down entire system of glassware. Element containers also act as spill containment chambers. If spill occurs, the whole container can be economically replaced without damage to other parts of the mantle.
All models feature clear anodized aluminum cabinet with black fluoropolymer resin-coated stainless steel top. Three-position units accommodate 500 to 1000 mL flasks. Six-position units accommodate 100 to 300 mL flasks. “RX” version mantles are used with solvents common to Soxhlet extractions, “RJ” version mantles are for refluxing aqueous solutions. Mantle dimensions, 3-position: 62L×31.7D×15.8H cm (241/2×121/2×61/4"); 6-position, 74L×26D×13.3H cm (291/4×101/2×51/4").
Two power control choices are available. Percentage timer version pulses full-line voltage to each heating position according to the dial setting. Proportional voltage version supplies a constant, steady-state voltage to each position. Both come equipped with quick-attach bracket for 1.2 cm (1/2") diameter rod, 1.8 m (6') multiconductor interconnect cord to mantle, and 1.2 m (4') long, three-wire power cord with grounding plug. The six-position timers and power controls are equipped with two brackets, two interconnect cords, and two power cords. Dimensions: 20.9H×11.4W×10.7D cm (81/4×41/2×41/4").
Clamps are used to hold the condensors/extraction glassware.
Ordering information: Mantles complete with heating element and a glassware support superstructure consisting of two upright rods, two horizontal rods, four lab clamps, and three spring-type glassware clamps that attach to one of the horizontal support rods. Heating mantle not equipped with power control; please order separately.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty