Would you like to log in again?
Hot Plates, Thermo Scientific
Supplier: Thermo Scientific
HPA2235MQ HPA2245MQ HPA2240MQ HPA2230MQ
33922-804EA 2600.57 USD InStock
33922-804 33922-859 33922-906 33922-757
Hot Plates, Thermo Scientific
Heaters Hot Plates
These hot plates feature demand-type thermostat controls that will automatically reach and hold temperature to within ±3 °C (5 °F) at 100 °C (212 °F).
  • Temperature range: 38 to 371 °C (100 to 700 °F)
  • Heavy, cast-aluminum top distributes heat evenly
  • Excellent for big heating jobs or multiple smaller ones
Plates feature stepless temperature selection, minimum overshoot, cycle light, and baffled air cooling. Heating element wires are embedded for protection, but are easy to replace if necessary.
Certifications: UL and C-UL listed.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty