Would you like to log in again?
Disrupter Horns and Microtips, Branson Ultrasonics
Supplier: Branson
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
101-148-013 101-148-063 101-148-070 101-063-212 101-147-039 101-147-047 101-148-069 101-063-212 101-147-038 101-148-013 101-147-035R 101-148-062 101-147-037R 101-148-062 101-147-050 101-148-069 101-147-048 101-148-063 101-148-070
33996-174EA 430.35 USD InStock
33996-174 33996-163 33996-185 CA33996-243 CA33996-265 33996-265 33996-243 CA33996-185 CA33996-163 CA33996-174 CA33996-108 33996-108 33995-324 33995-335 33995-334 33995-323 33995-325 33995-336 33995-330
Disrupter Horns and Microtips, Branson Ultrasonics
Homogenizers Ultrasonic Cell Disruptors
Disrupter Horns and Microtips for use with Sonifier® cell disrupters (see 33995-590 series).
  • All horns and tips are titanium
  • Horns transmit ultrasonic energy into a solution
  • The dimensions of the horn and the output control setting determine the amount of amplitude and degree of ultrasonic activity in the liquid
A step horn with a threaded body enables the attachment of various screw-on accessories, such as a continuous flow cell
Flat tips are ideal for general processing applications. Tapered micro tips are for processing small volumes (1 to 10 ml) and attach directly to the 13 mm (1/2") tapped disrupter horn 33995-323. The double stepped microtip assembly consists of a coupler and a stepped tip and attaches directly without need for a tapped horn. It has lower amplitude than the tapered tip, but is capable of reaching into small diameter vessels of greater depth and can process solutions ≤1 µL.
Tapped horn ends enable attachment of microtips. Tapped horns are supplied with removable flat tips. High gain horns cannot be used with screw-on accessories but provide higher amplitude at 19 mm (3/4") and 25 mm (1") diameters than the same diameter step horn. Exponential horns have lower tip amplitude, can withstand higher tip loading, and are best suited for applications requiring the compressing of solids such as powders and tissue. Cup horns enable sonification of a sample without the horn coming in contact with the solution. They can simultaneously process a number of sealed vials or test tubes with identical parameters.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty