Would you like to log in again?
Universal Grinding Mills, IKA® Works
Supplier: IKA
0001603603 1603600
33998-048EA 9304.61 USD InStock
33998-048 33998-050
Universal Grinding Mills, IKA® Works
Mills
Suitable for grinding up to 250mL of hard, brittle substances
Maximum grain size is 6 to 7mm. Unit features an exchangeable grinding chamber with handles. Exchangeable cutters for universal dry crushing. Easy to clean. Cast aluminum housing. Output: 225 watts.
Ordering information: Supplied with stainless steel blade, grinding chamber and lid, cord, and plug.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty