Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Orbital Shakers, 230 V, KS 260

Supplier: IKA

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

2980200 2980300
33998-362EA 5307.96 USD InStock
33998-362 33998-358
Orbital Shakers, 230 V, KS 260
Shakers and Mixers Orbital Shakers
For a variety of applications including medical diagnostics, hematology, water and wastewater testing, and dairy farming

  • 230 V model

Basic model features electronic adjustment of speed, and an LED line speed display. Control model features a digital display for speed, timer function, and operating modes. All functions of the Control model can be controlled and documented with labworldsoft* software via an integral RS-232 interface.

The universal platform holds various types of vessels and features variable clamping rolls that allow for universal adaptation. Universal platform surface dimensions: 32×32 cm (12⁹/₁₆×12⁹/₁₆"). The flask clamp platform is for shaking flasks, Erlenmeyer flasks, and bottles with a round cross-section. Accommodates fifty-six 25 ml flasks, twenty-three 50 or 100 ml flasks, eleven 200 or 250 ml flasks, or nine 500 ml flasks. The dish attachment is for smooth movement of cell cultures, nutrient media, petri dishes, culture bottles, and vessels with a low center of gravity. Dish platform surface dimensions: 32×32 cm (12⁹/₁₆×12⁹/₁₆").

May be used inside refrigerators or incubators within the specified temperature range. Features include a 50 W motor, 10 mm (²/₈") orbit diameter, and an electronic time switching clock. Clock can be set for timed or continuous operation.

Ordering information: Control models include labworldsoft software, a 2.5 m (8.2') PC 1.5 cable, and a PC 2.2 adapter. Universal platform includes four clamping rolls and eight fastening screws.

Caution: 230 V models. Corresponding models with voltage suitable for use in US (733998-360, 33998-364) are also available, please see separate product page.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty