Would you like to log in again?
SP Bel-Art Electrode Rack, Bel-Art Products, a part of SP
Supplier: Bel-Art
SCIENCEWARE®
H18509-0000
34102-089EA 124.01 USD InStock
34102-089
SP Bel-Art Electrode Rack, Bel-Art Products, a part of SP
Electrode Holders
Handy rack holds 8 test tubes for storing and conditioning pH and ion electrodes.
  • Polypropylene construction
  • Suitable for storing and conditioning electrodes
  • Chemically resistant
  • Autoclavable
The polypropylene rack resists chemical attack and is autoclavable at 121 °C (250 °F). Rack accomodates two rows of 4 tubes, up to 20 mm in diameter.
LxWxH: 10.8 x 6 x 12.1 cm
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty