Would you like to log in again?
Orion™ Stainless-Steel Automatic Temperature Compensation (ATC) Probes, Thermo Scientific
Supplier: Thermo Scientific
Orion™
928007MD 927007MD 927007 917007 928007MD 917004
89031-456EA 679.55 USD InStock
89031-456 34104-256 CA89031-456 14004-292 34104-258 10010-744
Orion™ Stainless-Steel Automatic Temperature Compensation (ATC) Probes, Thermo Scientific
Electrodes pH Electrodes
Automatically correct pH measurements for variation in electrode slope due to temperature change using Orion™ Stainless-Steel Automatic Temperature Compensation (ATC) Probes.
  • pH range of 0 to 14
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty