Would you like to log in again?
Orion™ pH Electrode Accessories, Thermo Scientific
Supplier: Thermo Scientific
Orion™
910003 910004-WA 910005
34104-723PK 145.53 USD InStock
34104-723 10010-632 34104-560
Orion™ pH Electrode Accessories, Thermo Scientific
Bottles Plastic Bottles
Stores and protects Orion™ pH electrodes.
  • Electrode storage bottles can be used with any 6, 8, or 12 mm pH electrodes
  • Electrode bulb guards are designed for use with glass body pH electrodes
Storage bottles protect and condition electrodes when not in use, and bulb guards protect the pH sensing bulb of glass pH electrodes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty