Would you like to log in again?
Orion™ Shorting Strap for BNC Meters, Thermo Scientific
Supplier: Thermo Scientific
Orion™
90045
34104-823EA 107.95 USD InStock
34104-823
Orion™ Shorting Strap for BNC Meters, Thermo Scientific
Cables
Shorting strap for use on pH meters with BNC input to perform BNC input verification testing and meter self test.
The strap may be used with pH meters with BNC input (Orion™ Versa Star™ pH, pH/ISE, and pH/LogR meters; Orion™ Star A series pH, pH/ISE, pH/conductivity, pH/RDO/DO, and pH/ISE/conductivity/RDO/DO meters; Orion™ Dual Star pH/ISE meters; Orion™ 2-Star, 3-Star, 4-Star and 5-Star pH meters).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty