Would you like to log in again?
Orion™ ROSS™ Combination pH Electrode, Thermo Scientific
Supplier: Thermo Scientific
Orion™
8102SC 8102BN
34107-733EA 1250.11 USD InStock
34107-733 34107-399
Orion™ ROSS™ Combination pH Electrode, Thermo Scientific
Electrodes pH Electrodes
General purpose, top performance combination electrode.
  • Works in extreme case of samples varying from one another by 50 °C or more
  • pH range of 0 to 14
  • Drift is less than 0.002 pH per day; restandardization is minimized
  • Double junction
  • Tris compatible
  • Waterproof BNC connector
Get stable readings to 0.01 pH in less than 30 s with the Orion™ ROSS™Combination pH Electrode. This electrode provides results three to five times more precise than those obtained with conventional electrodes. Recommended for Q.C. and research applications. Includes one 60 ml bottle of fill solution (3 M KCl) and spout cap.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty