Would you like to log in again?
Orion™ ROSS™ Combination Flat Surface pH Electrode, Thermo Scientific
Supplier: Thermo Scientific
Orion™
8135SC 8135BN
34107-737EA 1385.38 USD InStock
34107-737 34107-650
Orion™ ROSS™ Combination Flat Surface pH Electrode, Thermo Scientific
Electrodes pH Electrodes
Stable readings to 0.01 pH in less than 30 s.
  • pH range of 0 to 14
  • Glass fiber junction
  • pH precision: 0.01
The Orion™ ROSS™ Combination Flat Surface pH Electrode measures pH of soft, moist surfaces; both solid and semi-solid. This includes agar gel plates, meats, bread dough, and similar samples. With drift less than 0.002 pH per day and minimized restandardization, this electrode works in extreme cases of samples varying from one another by 50 °C or more. Includes one bottle of 3 M KCl filling solution.
Get results three to five times more precise than those obtained with conventional electrodes with the Orion™ ROSS™ Combination Flat Surface pH Electrode.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty