Would you like to log in again?
Orion™ Economy Series pH Combination Electrodes, Thermo Scientific
Supplier: Thermo Scientific
913600 9106BNWP 911600
34107-978EA 441.31 USD InStock
34107-978 34107-912 10010-786
Orion™ Economy Series pH Combination Electrodes, Thermo Scientific
Electrodes pH Electrodes
Conduct routine pH measurements with the Orion™ Economy Series pH Combination Electrode, a routine wick junction, gel-filled electrode.
  • Unbreakable epoxy body extends beyond the pH sensing glass bulb for rugged conditions
  • Provides durability for laboratory or field use with a low-maintenance design that requires no fill solution
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty