Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Orion™ Dissolved Oxygen/BOD Probe, Thermo Scientific
Supplier: Thermo Scientific
Orion™
970899WP 970803 970802 970801 970899WP
10010-828EA 4053.56 USD InStock
10010-828 34111-294 34111-272 CA10010-828 34111-308
Orion™ Dissolved Oxygen/BOD Probe, Thermo Scientific
Electrodes Dissolved Oxygen Electrodes
Make any pH meter a dissolved oxygen meter with the Orion™ Dissolved Oxygen/BOD Probe.
  • Polarographic O₂ sensor plus solid-state circuitry in probe top; converts current to voltage for direct DO readout
  • Plug probe into benchtop or portable pH meter, read DO over 0–14 ppm range
  • Accuracy at 15 to 35 °C (59 to 95 °F): ±0.05 ppm/2% of reading, whichever is greater
  • Dual thermocompensator corrects for membrane permeability and O₂ solubility
  • Ideal for BOD testing; overflow funnel supplied fits 300 ml BOD bottle; built-in magnetic stir bar
  • Screw-on membrane/electrolyte module
Accessories information: Membrane caps reduces probe servicing time and eliminate assembly problems using Orion™ Membrane Caps for Polarographic Dissolved Oxygen Probe. Convenient screw-on membrane caps help ensure optimal probe performance.
Ordering information: Electrode operates on two Mallory® type 10L14 batteries or equivalent. Supplied with two membrane/electrolyte modules, one overflow funnel, and batteries.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty