Would you like to log in again?
Orion™ ROSS™ Combination Semi-Micro pH Electrode, Thermo Scientific
Supplier: Thermo Scientific
Orion™
8103SC 8103BN
34107-734EA 994.6 USD InStock
34107-734 34116-061
Orion™ ROSS™ Combination Semi-Micro pH Electrode, Thermo Scientific
Electrodes pH Electrodes
Receive stable readings to 0.01 pH in less than 30 s with the Orion™ ROSS™ Combination Semi-Micro pH Electrode.
  • Works in extreme case of samples varying from one another by 50 °C or more
  • Drift is less than 0.002 pH per day; restandardization is minimized
  • Fits test tubes and measures samples as small as 0.2 ml
  • pH range of 0 to 14
  • pH precision: 0.01
  • Semi-micro tip
Orion™ ROSS™ Combination Semi-Micro pH Electrodes are recommended for use in clinical, pharmaceutical, and food laboratories, wherever sample size is a constraint. Includes one 60 ml bottle of 3 M KCl filling solution.
Results are three to five times more precise than those obtained with conventional electrodes.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty