Would you like to log in again?
Orion™ Fluoride Electrodes, Thermo Scientific
Supplier: Thermo Scientific
Orion™
9609BNWP 9409BN 9409SC 9609BNWP
10010-808EA 1388.51 USD InStock
10010-808 CA10010-808 34117-010 34117-932
Orion™ Fluoride Electrodes, Thermo Scientific
Electrodes ISE Electrodes
Quickly, simply, accurately, and economically measure free fluoride ions in aqueous solutions with the combination Orion™ Fluoride Electrodes.
Measure free fluoride ions in aqueous solutions with the Orion™ Fluoride Electrodes. The Sure-Flow™ junction allows easy cleaning and is easy to use in tough samples. The free flowing Sure-Flow™ junction assures stable, drift-free potentials. The half cell electrode requires a separate reference electrode. The reference electrode 900100 is recommended for use with thalf cell electrodes 9409BN and 9409SC.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty