Would you like to log in again?
Orion™ ISE Filling Solutions, Thermo Scientific
Supplier: Thermo Scientific
Orion™
900065 900067 900018-WA 900062 900017 900010 951202 900012 900063 900061 951209 900046 951209
15023-338PK 169.72 USD InStock
15023-338 34117-967 34117-965 34184-609 34109-157 34117-969 34113-948 CA97008-306 34117-963 34117-961 34108-022 34108-026 97008-306
Orion™ ISE Filling Solutions, Thermo Scientific
Electrolytes Electrode Filling Solutions
Enhance the performance and extend the life of ion selective electrodes with Orion™ ISE Filling Solutions.
  • Filling/maintenance solutions are for use with Orion™ Ion Selective Electrodes (ISE)
  • Optimum Result™ ISE filling solutions are specially designed for the Ionplus™ series ion selective electrodes
These solutions are specially formulated to work with the recommended ion selective electrodes and prevent electrode interferences from filling solution.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty