Would you like to log in again?
Orion™ Nitrate Electrodes, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Orion™
930701 930702 970701 9707BNWP 9707BNWP
34112-581PK 623.83 USD InStock
34112-581 21000-046 34112-586 34118-106 CA21000-046
Orion™ Nitrate Electrodes, Thermo Scientific
Electrodes ISE Electrodes
Measure nitrate in aqueous solutions simply, accurately, and economically with Orion™ Nitrate Electrodes.
  • Drift-free potentials
  • Free-flowing Sure-Flow junction
A separate reference electrode is unnecessary, making these electrodes convenient to use with small sample volumes.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty