Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nitric acid 69%, Veritas®, redistilled

Supplier: GFS CHEMICALS INC
Danger
Synonyms: Nitric acid solution

63-53105 53102
34126-320EA 279.25 USD InStock
34126-320 34126-324
Nitric acid 69%, Veritas®, redistilled
Nitric acid


Formula: HNO₃
MW: 63.01 g/mol
Boiling Pt: 120.5 °C (1013 hPa)
Melting Pt: –42 °C
Density: 1.4 g/cm³ (20 °C)
MDL Number: MFCD00011349
CAS Number: 7697-37-2
UN: 2031
ADR: 8,II
Order Now
Specification Test Results
Assay Min. 68.0%
Chloride 0.00001%
Sulfate 0.00001%
Arsenic 0.0000001%
Heavy Metals 0.000001%
Iron 0.000001%
Residue after Ignition 0.0001%
Color Max. 10 APHA

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty