Would you like to log in again?
pH Buffer Capsules, Hydrion®
Supplier: Micro Essential Laboratory
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
270-10.00 270-2.40 270-12.00 270-11.00 270-9.00 270-7.40 270-6.80 270-5.00 270-3.00 270-8.40 270-8.00 270-7.20 270-2.00 270-4.00 270-7.00
34175-071CS 69.1 USD InStock
34175-071 34175-173 34175-480 34175-129 34175-231 34175-297 34175-242 34175-435 34175-049 34175-027 34175-399 34175-377 34175-300 34175-344 34175-322
pH Buffer Capsules, Hydrion®
Buffers pH Calibration Buffers
For the quick and convenient preparation of fresh solutions of known pH values
Capsule contents dissolve in 100mL of distilled water, giving a clear solution accurate to ±0.02 pH unit. The dry powder remains stable. Each vial contains 10 capsules.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty