Would you like to log in again?
Hydrion pH Buffers, Micro Essential Lab
Supplier: Micro Essential Laboratory
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Hydrion®
2807400 2801100 280200 280600 280-3 280800 280500 280686 280640 280700 280900 2801000 CC-725 2801200 280400
34175-093PK 236.28 USD InStock
34175-093 34175-081 34175-091 34175-097 34175-095 34175-083 60785-007 62000-406 34175-079 34175-089 34175-099 60775-984 34175-077 34175-087 60775-986
Hydrion pH Buffers, Micro Essential Lab
Buffers pH Calibration Buffers
Chemvelopes are made of a triple protective foil consisting of aluminum, polyethylene and paper which keeps the Hydrion Buffers dry and free-flowing.
Hydrion buffer chemvelopes produce 500 ml of pH buffer standard solution when dissolved in distilled water.
Packaging: Each carton contains five chemvelopes.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty