Would you like to log in again?
Orion™ pH Buffer Individual Use Pouches, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
ORION®
910710 910725 911010 910410-WA 911025-WA 910425
34175-514PK 60.65 USD InStock
34175-514 34175-518 34175-512 34175-522 34175-510 34175-520
Orion™ pH Buffer Individual Use Pouches, Thermo Scientific
Buffers pH Calibration Buffers
Ensure fresh, ready to use standards for pH calibration with Orion™ pH Buffer Individual Use Pouches
  • Color coded: pH 4.01 buffer is pink, pH 7.00 buffer is yellow, and pH 10.01 buffer is blue
  • Packaged with buffer/temperature tables for accurate calibrations and NIST traceability
  • Manufactured under ISO 9000 quality standards
  • Pouches are 15mL each and are available in 10 packs or 25 packs
Each pouch is sealed against light, air, or other contamination, and is disposable. There is no wasted buffer or secondary reservoir. Each pouch contains lot code, expiration date, and buffer temperature table.
Buffers are color coded for ease of use. All Orion™ pH buffers are traceable to NIST standard reference materials.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty