Would you like to log in again?
Orion™ Total Alkalinity Test Kit, Thermo Scientific
Supplier: Thermo Scientific
700011-WA 700012 700010
34182-060EA 412.98 USD InStock
34182-060 23198-342 34182-064
Orion™ Total Alkalinity Test Kit, Thermo Scientific
Environmental Test Kits
Save time by eliminating additional equipment set-up, titrations, and calculations
  • Direct reading of total alkalinity from 0 to 200ppm as CaCO3, from the enclosed pH conversion chart
  • No interference from color or turbidity
The Orion™ Total Alkalinity Test Kit provides two-step direct measurement of total alkalinity and pH, using a pH electrode.
Ordering information: Test kits include one 475mL bottle of alkalinity reagent, one 475mL bottle of alkalinity standard/control, total alkalinity conversion chart, and application procedure.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty