Would you like to log in again?
Orion™ pH Buffer Bottles, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
ORION®
910686 9116860 910760 9191860 910104 910686 910112 910168 9116860 910918 916860 910110 910112 9191860 910460 911260-WA 916860 910168 910918 911060 910105 910107
34182-306EA 46.72 USD InStock
34182-306 CA10010-640 10010-642 10010-640 CA10010-634 10010-646 CA10010-646 10010-634 10010-644 CA10010-636 23198-220 CA10010-648 10010-638 23198-222 10010-648 10010-636 CA10010-644 23198-224 34182-390 CA10010-638 34182-310 34182-386
Orion™ pH Buffer Bottles, Thermo Scientific
Buffers pH Calibration Buffers
Perform accurate and reproducible pH calibrations with Orion™ pH buffer bottles.
  • All bottles feature pH/temperature tables printed on the labels for accurate calibrations and NIST traceability
  • Manufactured under ISO 9000 quality standards
  • Available in pint (475 ml) bottles or portable five packs of 2 oz. (5×60 ml) bottles
The pH 4.01, 5.00, 7.00, and 10.01 buffers are color-coded for ease of use. All Orion™ pH buffers are traceable to NIST standard reference materials.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty