Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 25, 2024. During this time, our site will be not available. We appreciate your patience.

VWR® Baume Plain Form Hydrometers, Traceable to NIST

Supplier: VWR
VWR®
61804-0011 61804-0111 61804-0411 61804-0711 61804-0911 61804-0211 61804-1011 61804-0511 61804-0811 61804-0611 61804-0311
34580-003EA 45.2 USD InStock
34580-003 34585-007 34559-903 34559-925 34559-947 34559-969 34565-000 34575-003 34570-000 34559-980 34560-006
VWR® Baume Plain Form Hydrometers, Traceable to NIST
Hydrometers
Available in a variety of models measuring Baume of syrups and other liquids heavier than water in various ranges from 0° to 70°, these plain form hydrometers are used in the chemistry, pharmacology, food and beverage, starch industries and more.

  • Lead free ballast and glass
  • Individually serialized
  • Temperature standardization: 60 °F (15 °C)
  • Hand inspected by our U.S. quality team

Certifications: Traceable to NIST.

Packaging: Packaged with a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy.

Caution: Hydrometers should be stored in a clean, dry, safe place, and should not be subjected to extreme temperatures.

Order Now
 

ORDER

SERVICES


Services: VWR CATALYST™ provides convenient and efficient re-calibration services for your hydrometers with an industry leading 5-day average turn-around time from receipt of your instrument. All the service will be performed by an ISO17025 certified lab. Recalibration, certification and shipping fees are all included in the catalog number cost. Simply order the stated catalog number based on how many calibration points are desired. You will receive a protective box to ship your instrument to Avantor's certified calibration laboratory along with a work order form to specify your calibration points and prepaid return shipping label. Upon receipt at the laboratory your instrument will be inspected for any defects under a microscope and the performance of your instrument will be compared to certified standards and corrections to one-tenth of the smallest division recorded on your signed calibration report. Your calibration report and instrument are carefully repacked returned to you with prepaid standard ground shipping. We also provide expedited service options available upon request.

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty