Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® Specific Gravity/Relative Density (g/cm³) Precision Hydrometers, Traceable to NIST

Supplier: VWR

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

VWR®
61801-0711 61801-0011 61801-0111 61801-2111 61801-1511 61801-0511 61801-1411 61801-2011 61801-1011 61801-1711 61801-1111 61801-0311 61801-1811 61801-0811 61801-0211 61801-1611 61801-1311 61801-0911 61801-1211 61801-0411 61801-0611
34627-650EA 62.48 USD InStock
34627-650 34627-560 34627-231 34627-253 34627-297 34627-173 34627-130 34627-174 34627-050 34627-082 34627-628 34627-537 34627-504 34627-118 34627-027 34627-479 34627-446 34627-413 34627-311 34627-355 34627-388
VWR® Specific Gravity/Relative Density (g/cm³) Precision Hydrometers, Traceable to NIST
Hydrometers
Available in a wide range of models measuring specific gravity/relative density (g/cmᶟ) of a variety of liquids in various ranges from 0.600 to 1.920, with 0.0005 divisions.

  • 325 mm (12.8”) long
  • Lead free ballast and glass
  • Individually serialized
  • Temperature standardization: 60°F (15°C)
  • Hand inspected by our U.S. quality team

Specific gravity hydrometers are the most widely used hydrometer, common in industries such as food and beverage, chemical, pharmacology, transportation, refrigeration and more.

Packaging: Packaged with a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy indicating accuracy traceable to NIST.

Order Now
 

ORDER

SPECIFICATIONS

SERVICES


Services: VWR CATALYST™ provides convenient and efficient re-calibration services for your hydrometers with an industry leading 5-day average turn-around time from receipt of your instrument. All the service will be performed by an ISO17025 certified lab. Recalibration, certification and shipping fees are all included in the catalog number cost. Simply order the stated catalog number based on how many calibration points are desired. You will receive a protective box to ship your instrument to Avantor's certified calibration laboratory along with a work order form to specify your calibration points and prepaid return shipping label. Upon receipt at the laboratory your instrument will be inspected for any defects under a microscope and the performance of your instrument will be compared to certified standards and corrections to one-tenth of the smallest division recorded on your signed calibration report. Your calibration report and instrument are carefully repacked returned to you with prepaid standard ground shipping. We also provide expedited service options available upon request.

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty