Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
SP Bel-Art Hydrometer Storage Racks, Bel-Art Products, a part of SP
Supplier: Bel-Art
Scienceware®
F18988-0000
34629-990CS 1099.27 USD InStock
34629-990
SP Bel-Art Hydrometer Storage Racks, Bel-Art Products, a part of SP
Racks Hydrometer Racks
Protect fragile glass hydrometers by storing them in this 18-place polypropylene rack.
  • Polypropylene construction
  • Broad base helps prevent tipping
  • Autoclavable
The middle and top plates feature 25 mm (1") diameter holes. The bottom plate is 44 mm (1¾") high and features wells tapering from 25 mm to 9 mm (1 to ⅜") to hold the hydrometer bulb. The base is 21x21.6 cm (8¼x8½") for tip-proof stability. The rack's overall height is 25.1 cm (9⅞") with handles. Steam autoclavable at 121°C (250°F).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty