Would you like to log in again?
Combined Form, Mercury-Free Hydrometers, API, G&W Instruments
Supplier: G&W Instruments
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
SA-913L SA-912L SA-918L SA-905L SA-907L SA-972L SA-975L SA-974L SA-919L SA-910L SA-908L SA-916L SA-921L SA-903L SA-904L SA-906L SA-976L SA-911L SA-973L SA-902L SA-909L SA-920L
34635-040EA 79.39 USD InStock
34635-040 34635-004 34635-048 34635-016 34635-038 34635-046 34635-002 34635-014 34635-036 34635-022 34635-044 34635-000 34635-034 34635-012 34635-020 34635-042 34635-032 34635-010 34635-008 34635-006 34635-028 34635-018
Combined Form, Mercury-Free Hydrometers, API, G&W Instruments
Hydrometers
These petroleum hydrometers have a thermometer in the body
Thermometers are filled with organic red liquid and contain no mercury.
They measures specific gravity according to API degree scale. Approximate length: 380mm. Thermometer range: 0 to 150°F. Temperature of standardization: 60°F.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty