Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® Specific Gravity/Relative Density and Baume Plain Form Hydrometers, Traceable to NIST

Supplier: VWR

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

VWR®
61806-0411 61806-0211 61806-0111 61806-1011 61806-1111 61806-0911 61806-0511 61806-0311 61806-1211 61806-0811
34640-320EA 48 USD InStock
34640-320 34640-342 34640-309 34640-284 34640-262 34640-207 34670-128 34670-106 34670-140 34670-161
VWR® Specific Gravity/Relative Density and Baume Plain Form Hydrometers, Traceable to NIST
Hydrometers
Used in the chemistry, pharmacology, food and beverage, and starch industries to name a few, these dual scale, plain form hydrometers measure specific gravity / relative density (g/cm3) of liquids lighter and heavier than water in various ranges from 0.700 to 2.00, and Baume in various ranges from 10° to 70°

  • Lead free ballast and glass
  • Individually serialized
  • Temperature standardization: of 60 °F (15 °C)
  • Hand inspected by our U.S. quality team

Certifications: Traceable to NIST.

Delivery information: Packaged with a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy.

Caution: Hydrometers should be stored in a clean, dry, safe place, and should not be subjected to extreme temperatures.

Order Now
 

ORDER

SERVICES


Services: VWR CATALYST™ provides convenient and efficient re-calibration services for your hydrometers with an industry leading 5-day average turn-around time from receipt of your instrument. All the service will be performed by an ISO17025 certified lab. Recalibration, certification and shipping fees are all included in the catalog number cost. Simply order the stated catalog number based on how many calibration points are desired. You will receive a protective box to ship your instrument to Avantor's certified calibration laboratory along with a work order form to specify your calibration points and prepaid return shipping label. Upon receipt at the laboratory your instrument will be inspected for any defects under a microscope and the performance of your instrument will be compared to certified standards and corrections to one-tenth of the smallest division recorded on your signed calibration report. Your calibration report and instrument are carefully repacked returned to you with prepaid standard ground shipping. We also provide expedited service options available upon request.

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty