Would you like to log in again?
VWR® Combined Alcohol Proof and Tralles Hydrometer
Supplier: VWR
VWR®
61807-8011
34715-000EA 93.87 USD InStock
34715-000
VWR® Combined Alcohol Proof and Tralles Hydrometer
Hydrometers
Dual scale hydrometer features thermometer in the bottom portion to permit temperature correction of test liquid.
  • Proof range of 0 to 200
  • Tralles range of 0 to 100
  • Temperature standard: 60°F
Certifications: Each hydrometer is calibrated in a triple accredited ISO/IEC 17025:2005, A2LA accredited laboratory against equipment whose calibration is traceable to NIST.
Packaging: Individually packaged with a multi-language Statement of Accuracy indicating accuracy traceable to NIST.
Order Now
ORDER
SPECIFICATIONS
SERVICES
Services: VWR CATALYST™ provides convenient and efficient re-calibration services for your hydrometers with an industry leading 5-day average turn-around time from receipt of your instrument. All the service will be performed by an ISO17025 certified lab. Recalibration, certification and shipping fees are all included in the catalog number cost. Simply order the stated catalog number based on how many calibration points are desired. You will receive a protective box to ship your instrument to Avantor's certified calibration laboratory along with a work order form to specify your calibration points and prepaid return shipping label. Upon receipt at the laboratory your instrument will be inspected for any defects under a microscope and the performance of your instrument will be compared to certified standards and corrections to one-tenth of the smallest division recorded on your signed calibration report. Your calibration report and instrument are carefully repacked returned to you with prepaid standard ground shipping. We also provide expedited service options available upon request.
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty