Would you like to log in again?
SRK® Collection and Transport System 922C, COPAN Diagnostics
Supplier: COPAN Diagnostics
SRK™
922C
35001-166CS 580.03 USD InStock
35001-166
SRK® Collection and Transport System 922C, COPAN Diagnostics
Swabs Transport Swabs
COPAN’s SRK® system combines a swab with a rinse solution or broth in a convenient plastic tube.
  • 10 cm traditional polyester swab with skirted tube containing 10 ml COPAN SRK® solution
  • With plastic green capture cap
  • Individually wrapped, sterile
The environmental sampling line helps food technologists and quality control managers assess microbial presence on surfaces and equipment.
Delivery information: 500 pcs per carton (10 boxes of 50 pcs).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty