Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
CRYOBANK RD CAP+BEADS PK80
Supplier: COPAN DIAGNOSTICS INC
CRYOBANK* Cap and Beads Red^ CRYOBANK* Cap and Beads, Color: Red, Chemically treated ceramic beads suspended in a special cryogenic preserving solution, The special cryogenic medium consists of a phosphate buffered saline supplemented with sucrose, glycerol and peptone, Reliable, convenient
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CRYOBANK RD CAP+BEADS PK80 35001-200 CRYO80C/R Box of 80 Retrieving
Pack of 50 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty