Would you like to log in again?
Lab dancer Vortex Mixers, IKA® Works
Supplier: IKA
0003365000
35006-032EA 1255.03 USD InStock
35006-032
Lab dancer Vortex Mixers, IKA® Works
Shakers and Mixers Vortex Mixers
Designed for mixing small test samples with a touch function.
  • Small, compact and reliable
  • For small containers up to 30 mm in diameter, e.g. test tubes, centrifuge tubes, Eppendorf beakers
  • With excellent mixing action
  • Includes light 12 V power pack set
The upper casing and the test tube surface are made from inert plastic. The bottom section is a coated, zinc die-casting.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty