Would you like to log in again?
VWR® Digital Hygrometers
Supplier: VWR
VWR®
61018-32 VC3003 90008-32 VC4096 61018-31
35519-046EA 154.46 USD InStock
35519-046 10754-540 35519-047 10754-726 35519-045
VWR® Digital Hygrometers
Hygrometers Digital Thermo-Hygrometers
Digital hygrometers are ideal for critical experiments, routine measurements, and continuous monitoring applications, and are suitable for all quality control needs.
  • Traceable®
  • Contain fast response solid-state sensors
  • Units perform measurements 2.5 times a second and update the display once a second
A 1.5 cm (5/8") high LCD display shows readings and low battery
Hygrometer 35519-045 and hygrometer/thermometer 35519-047 feature minimum/maximum memory. Minimum and maximum values may be displayed or cleared at any time with the touch of a button.
Hygrometers/thermometers 35519-046 and 35519-047 continuously and simultaneously display relative humidity and temperature. Temperature display can be switched from °F to °C.
Units run continuously for one year on one set of batteries. No adjustments or maintenance required. ABS case is chemical-resistant and shock-resistant.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology. Hygrometer/thermometer 35519-047 meets federal requirements for monitoring temperature and humidity in laboratories (CLIA specification 493.1252).
Ordering information: All units include a wall mount and two AAA 1.5V batteries. Units 35519-045 and 35519-047 also include a bench stand.
Order Now
ORDER
SPECIFICATIONS
SERVICES
Services: VWRCATALYST™ provides convenient and efficient re-calibration services for your digital hygrometers with an industry leading 5-day average turn-around time from receipt of your instrument. All the service will be performed by an ISO17025 certified lab. Recalibration, certification and shipping fees are all included in the catalog number cost. You will receive a protective box to ship your instrument to Avantor's certified calibration laboratory along with a work order form and a prepaid return shipping label. Upon receipt at the laboratory your instrument will be inspected for any defects and the performance of your instrument will be compared to certified standards. After re-calibration, your calibration report and instrument will be carefully repacked then returned with standard ground shipping. We also provide expedited service options available upon request. Hygrometer not included.
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty