Would you like to log in again?
Cole-Parmer® Dri-Block® BH-250 Heaters
Supplier: Antylia Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Dri-Block®
6014470 36620-59 36620-64 36620-55 36620-54 1082000 36620-65 36620-53 36620-15 36620-40 1100596 36620-57 36620-67 36620-13 36620-61 36620-97 36620-56 36620-11 36620-58 36620-96 36620-52 6014471 36620-99 36620-63 36620-60
35756-478EA 307.07 USD InStock
35756-478 35756-259 35756-578 35756-512 35756-476 35756-256 35756-510 35756-474 35756-508 35756-506 35756-504 14229-117 14229-010 35756-580 14229-012 35756-260 14228-922 35756-582 35756-518 14228-991 35756-438 14228-952 35756-516 35756-436 35756-514
Cole-Parmer® Dri-Block® BH-250 Heaters
Block Heaters Digital Block Heaters
Choose a heater, then blocks to suit your application needs.
  • Precision-engineered heaters with excellent temperature accuracy
  • Versatile range of interchangeable heating blocks work with various size sample tubes and plates
  • Take up less storage space - lightweight and flat-top surface allow units to be stacked when not in use
Use these heaters as a dry, constant temperature source for various lab and research applications. All models have independent indicator lights to show main power, heater action, and over-temperature cut out. The ‘number of blocks accepted’ on the ordering table indicate the number of single-width blocks accepted by each block heater. A heater that accepts two single-width blocks will accept one double-width block. Use three- and four-block models for high-throughput applications such as chemical oxygen demand (COD).
Techne® Dri-Block® heaters have earned a strong reputation for reliability. All instruments are supplied with a 3-year warranty. The anodised aluminium Dri-Block® inserts are supplied with a 1-year warranty.
Now Techne® are proud to introduce the new, updated range offering market-leading choice, flexibility, and specification. Dri-Block® heaters are a source of precision temperature control for general routing applications and sensitive analytical procedures particularly suited to microbiology and clinical laboratories. They are designed for precise heating of test-tubes, microcentrifuge tubes, cuvettes, and microtiter plates. Techne® Dri-Block® heaters cover a temperature range from ambient +5 to 100°C or 200°C (depending on model). These units use machined aluminium alloy Dri-Block® inserts as the heat transfer medium. Independent indicator lights indicate mains power, heater action, and over-temperature cut-out. An On/Off switch is mounted on the rear panel, together with the mains connector, a double fuse holder, and adjustable over-temperature cut-out (depending on model).
Ordering information: One pack of plastic spacer blocks (supplier no. F4466) is required for use with sample concentrator SC-4/96 Hood and Stand (supplier no. FSC496D), and must be ordered separately.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty