Would you like to log in again?
VWR SHELF F/2200 2250 5015/25
Supplier: VWR International
VWR* Stainless Steel Shelf^ Stainless steel shelf, 45x45cm, for use with VWR* Model 5015/5025 Air-Jacketed CO2 Incubators and VWR* Water-Jacketed CO2 Incubators, Models 2200/2250.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Unit Price Quantity
VWR SHELF F/2200 2250 5015/25 35910-886 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty