Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
VWR® Wide Mouth Round Bottles, HDPE, with Caps
Supplier: VWR
VWR®
PLC-03622 PLC-03611
16125-903CS 145.24 USD InStock
16125-903 36319-343
VWR® Wide Mouth Round Bottles, HDPE, with Caps
Bottles Water Sampling Bottles
Ideal for general laboratory use or for volatile organic analysis.
  • HDPE construction
  • Wide-mouthed design
  • Pre-cleaned in a contaminant-free environment
Precleaned bottles are washed in hot water and laboratory grade non-phosphate detergent, then rinsed three times with tap water and three times with ASTM Type I organic-free water.
They are finally oven dried and cooled to room temperature in an enclosed contaminant-free environment.
Ordering information: Supplied with F-217 polypropylene-lined caps. Pressure-sensitive sample labels are included in each carton.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty