Would you like to log in again?
Standard Wide Mouth Bottles, Vacuum and Ionized, Clear
Supplier: Qorpak
GLC-01759 GLC-01746 GLC-01803 GLC-01790 GLC-01756 GLC-01817 GLC-01787 GLC-01885 GLC-01861 GLC-01780 GLC-06457 GLC-01836 GLC-01869 GLC-01773
36319-794CS 115.11 USD InStock
36319-794 36319-793 11311-402 36319-790 TN7468C 36319-792 11311-478 36319-791 76487-370 89204-698 76487-374 76487-372 89204-696 36319-744
Standard Wide Mouth Bottles, Vacuum and Ionized, Clear
Bottles Glass Bottles
The vacuum and ionized cleaning process removes contaminants such as loose dirt, carton lint, aerosols, and fine glass particles.
  • Extremely cost-effective
  • No need to waste valuable time rewashing bottles
  • Provides a clean final appearance
  • Seals cleanliness into each bottle
  • Each container is individually cleaned, and will not collect dust, flies, and other debris while stored in the lab
Compressed air passes through a condensate separator and coalescing filter to remove aerosols and fine dust particles. Then the air passes through an absorbent filter to remove gases. A vacuum then draws the particulate-laden atmosphere out of the container. Individually cleaned closures are immediately placed on the container. The air inside the bottle will not interfere with the product in any way.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty